accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 93-52-7

BrBr

CAS RN 93-52-7

IUPAC Name: (1,2-dibromoethyl)benzene
Synonyms: (1,2-dibromoethyl)benzene
Molecular Weight (g/mol): 263.96
Molecular Formula: C8H8Br2
InChi Key: SHKKTLSDGJRCTR-UHFFFAOYNA-N
SMILES: BrCC(Br)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)