accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3O

CAS RN 93-53-8

IUPAC Name: 2-phenylpropanal
Synonyms: benzeneacetaldehyde, α-methyl-
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: IQVAERDLDAZARL-UHFFFAOYNA-N
SMILES: CC(C=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)