accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 93-89-0

IUPAC Name: ethyl benzoate
Synonyms: ethyl benzoate
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: MTZQAGJQAFMTAQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results