accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 93-99-2

IUPAC Name: phenyl benzoate
Synonyms: phenyl benzoate
Molecular Weight (g/mol): 198.22
Molecular Formula: C13H10O2
InChi Key: FCJSHPDYVMKCHI-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)