accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 930-30-3

IUPAC Name: cyclopent-2-en-1-one
Synonyms: cyclopentenone
Molecular Weight (g/mol): 82.1
Molecular Formula: C5H6O
InChi Key: BZKFMUIJRXWWQK-UHFFFAOYSA-N
SMILES: O=C1CCC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)