accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NN

CAS RN 930-36-9

IUPAC Name: 1-methyl-1H-pyrazole
Synonyms: 1-methyl-1H-pyrazole
Molecular Weight (g/mol): 82.11
Molecular Formula: C4H6N2
InChi Key: UQFQONCQIQEYPJ-UHFFFAOYSA-N
SMILES: CN1C=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results