accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOO

CAS RN 930-88-1

IUPAC Name: 1-methyl-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: N-methylmaleimide
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: SEEYREPSKCQBBF-UHFFFAOYSA-N
SMILES: CN1C(=O)C=CC1=O
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)