accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 930110-97-7

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 4-(oxan-4-yloxy)benzoate
Synonyms: 2,3,4,5,6-pentafluorophenyl 4-(oxan-4-yloxy)benzoate
Molecular Weight (g/mol): 388.29
Molecular Formula: C18H13F5O4
InChi Key: BDWCEFHYQPZQRO-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CC=C(OC3CCOCC3)C=C2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results