accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 93041-45-3

IUPAC Name: 3-(4-methoxyphenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Synonyms: 3-(4-methoxyphenyl)-5-methyl-1,2-oxazole-4-carboxylic acid
Molecular Weight (g/mol): 233.22
Molecular Formula: C12H11NO4
InChi Key: IKDPECCMAKOJTK-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C1=NOC(C)=C1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results