accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 931-03-3

HNOOH

CAS RN 931-03-3

IUPAC Name: 1H-pyrrole-3-carboxylic acid
Synonyms: 1H-pyrrole-3-carboxylic acid
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: DOYOPBSXEIZLRE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results