CAS RN 931-19-1
IUPAC Name:
2-methylpyridin-1-ium-1-olate
Synonyms:
picoline N-oxide methylpyridine 1-oxide
Molecular Weight (g/mol):
109.13
Molecular Formula:
C6H7NO
InChi Key:
CFZKDDTWZYUZKS-UHFFFAOYSA-N
SMILES:
CC1=CC=CC=[N+]1[O-]
1
–
1
of
1
results
1
2-Picoline N-oxide, 98%
CAS: 931-19-1 Molecular Formula: C6H7NO Molecular Weight (g/mol): 109.128 MDL Number: MFCD00006199 InChI Key: CFZKDDTWZYUZKS-UHFFFAOYSA-N Synonym: 2-picoline-n-oxide,2-methylpyridine 1-oxide,2-methylpyridine n-oxide,picoline n-oxide,2-picoline n-oxide,methylpyridine 1-oxide,alpha-picoline n-oxide,pyridine, 2-methyl-, 1-oxide,2-picoline, 1-oxide,picoline 1-oxide PubChem CID: 13602 ChEBI: CHEBI:35578 IUPAC Name: 2-methyl-1-oxidopyridin-1-ium SMILES: CC1=CC=CC=[N+]1[O-]