accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

MgCl

CAS RN 931-51-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 142.91
Molecular Formula: C6H11ClMg
InChi Key: DEDWWARPYWCXMG-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].C1CC[CH-]CC1
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)