accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 931-87-3

IUPAC Name: (Z)-cyclooctene
Synonyms: cyclooctene
Molecular Weight (g/mol): 110.2
Molecular Formula: C8H14
InChi Key: URYYVOIYTNXXBN-UPHRSURJSA-N
SMILES: C1CCC\C=C/CC1
Boiling Point 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)