accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 93107-30-3

IUPAC Name: 1-cyclopropyl-6,7-difluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: 1-cyclopropyl-6,7-difluoro-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol): 265.22
Molecular Formula: C13H9F2NO3
InChi Key: KNEXGVPHPGXAGF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN(C2CC2)C2=CC(F)=C(F)C=C2C1=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results