accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 932-30-9

IUPAC Name: 2-(aminomethyl)phenol
Synonyms: 2-(aminomethyl)phenol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: KPRZOPQOBJRYSW-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results