CAS RN 932750-29-3
CAS RN 932750-29-3
IUPAC Name:
ethyl 2-cyano-3-ethoxybut-2-enoate
Synonyms:
ethyl 2-cyano-3-ethoxybut-2-enoate
Molecular Weight (g/mol):
183.21
Molecular Formula:
C9H13NO3
InChi Key:
YKXFOGAYPIPTKF-UHFFFAOYSA-N
SMILES:
CCOC(C)=C(C#N)C(=O)OCC
1
–
1
of
1
results
1
Ethyl (E)-2-cyano-3-ethoxycrotonate, 98%
CAS: 932750-29-3 Molecular Formula: C9H13NO3 Molecular Weight (g/mol): 183.207 MDL Number: MFCD01743485 InChI Key: YKXFOGAYPIPTKF-BQYQJAHWSA-N Synonym: ethyl 2e-2-cyano-3-ethoxybut-2-enoate,ethyl e-2-cyano-3-ethoxycrotonate,ethyl 2-cyano-3-ethoxybut-2-enoate,2-butenoic acid, 2-cyano-3-ethoxy-, ethyl ester,ethyl e-2-cyano-3-ethoxybut-2-enoate,e-ethyl 2-cyano-3-ethoxybut-2-enoate,ethyl e-2-cyano-3-ethoxy-2-butenoate,e-2-cyano-3-ethoxycrotonic acid ethyl ester,e-2-cyano-3-ethoxy-2-butenoic acid ethyl ester PubChem CID: 6387109 IUPAC Name: ethyl (E)-2-cyano-3-ethoxybut-2-enoate SMILES: CCOC(=C(C#N)C(=O)OCC)C