accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 934-05-4

CH3OONHBr

CAS RN 934-05-4

IUPAC Name: methyl 4-bromo-1H-pyrrole-2-carboxylate
Synonyms: methyl 4-bromo-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 204.02
Molecular Formula: C6H6BrNO2
InChi Key: ZXFCRVGOHJHZNF-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(Br)=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results