accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NHNN

CAS RN 934-32-7

IUPAC Name: 1H-1,3-benzodiazol-2-amine
Synonyms: 2-aminobenzimidazole 2-iminobenzimidazoline
Molecular Weight (g/mol): 133.15
Molecular Formula: C7H7N3
InChi Key: JWYUFVNJZUSCSM-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=CC=C2N1
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results