accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 934-48-5

CH3CH3NNNH2O

CAS RN 934-48-5

IUPAC Name: 3,5-dimethyl-1H-pyrazole-1-carboxamide
Synonyms: 3,5-dimethylpyrazole-1-carboxamide
Molecular Weight (g/mol): 139.16
Molecular Formula: C6H9N3O
InChi Key: AKRDSDDYNMVKCX-UHFFFAOYSA-N
SMILES: CC1=NN(C(N)=O)C(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results