accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NOHO

CAS RN 934-60-1

IUPAC Name: 6-methylpyridine-2-carboxylic acid
Synonyms: 6-methylpyridine-2-carboxylic acid
Molecular Weight (g/mol): 137.14
Molecular Formula: C7H7NO2
InChi Key: LTUUGSGSUZRPRV-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=N1)C(O)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)