accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OS

CAS RN 934-87-2

IUPAC Name: 1-(phenylsulfanyl)ethan-1-one
Synonyms: thiacetic acid, S-phenyl ester
Molecular Weight (g/mol): 152.21
Molecular Formula: C8H8OS
InChi Key: WBISVCLTLBMTDS-UHFFFAOYSA-N
SMILES: CC(=O)SC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results