accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNNH2

CAS RN 934-98-5

IUPAC Name: 2-(4-methylpiperazin-1-yl)ethan-1-amine
Synonyms: 2-(4-methylpiperazin-1-yl)ethanamine
Molecular Weight (g/mol): 143.23
Molecular Formula: C7H17N3
InChi Key: GOWUDHPKGOIDIX-UHFFFAOYSA-N
SMILES: CN1CCN(CCN)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results