accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 934570-43-1

BOOCH3CH3CH3CH3NH3C

CAS RN 934570-43-1

IUPAC Name: 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-indole
Synonyms: 1-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydroindole
Molecular Weight (g/mol): 259.16
Molecular Formula: C15H22BNO2
InChi Key: CZYIGZLFKRAXMV-UHFFFAOYSA-N
SMILES: CN1CCC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results