accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHSN

CAS RN 934570-47-5

IUPAC Name: methyl({[5-(pyridin-4-yl)thiophen-2-yl]methyl})amine
Synonyms: methyl({[5-(pyridin-4-yl)thiophen-2-yl]methyl})amine
Molecular Weight (g/mol): 204.29
Molecular Formula: C11H12N2S
InChi Key: JSAZQLBHWDPNPA-UHFFFAOYSA-N
SMILES: CNCC1=CC=C(S1)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results