accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 934570-57-7

IUPAC Name: ({1-[(furan-2-yl)methyl]piperidin-4-yl}methyl)(methyl)amine
Synonyms: {[1-(furan-2-ylmethyl)piperidin-4-yl]methyl}(methyl)amine
Molecular Weight (g/mol): 208.31
Molecular Formula: C12H20N2O
InChi Key: LISJAFHYRUEIGV-UHFFFAOYSA-N
SMILES: CNCC1CCN(CC2=CC=CO2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results