CAS RN 935-14-8
IUPAC Name:
1,4-diethynylbenzene
Synonyms:
1,4-diethynylbenzene
Molecular Weight (g/mol):
126.16
Molecular Formula:
C10H6
InChi Key:
MVLGANVFCMOJHR-UHFFFAOYSA-N
SMILES:
C#CC1=CC=C(C=C1)C#C
1
–
1
of
1
results
1
1,4-Diethynylbenzene, 95%
CAS: 935-14-8 Molecular Formula: C10H6 Molecular Weight (g/mol): 126.16 MDL Number: MFCD00078375 InChI Key: MVLGANVFCMOJHR-UHFFFAOYSA-N Synonym: p-diethynylbenzene,benzene, 1,4-diethynyl,1,4-diethynyl-benzene,benzene, diethynyl,benzene,1,4-diethynyl,benzene, p-diethynyl-7ci,8ci,1,4-bis ethynyl benzene,acmc-209rm8,4-05-00-01805 beilstein handbook reference PubChem CID: 120463 IUPAC Name: 1,4-diethynylbenzene SMILES: C#CC1=CC=C(C=C1)C#C