accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 935-42-2

IUPAC Name: 1-(1-phenylcyclopropyl)methanamine
Synonyms: 1-(1-phenylcyclopropyl)methanamine
Molecular Weight (g/mol): 147.22
Molecular Formula: C10H13N
InChi Key: FUEMGCALFIHXAS-UHFFFAOYSA-N
SMILES: NCC1(CC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results