accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 935-44-4

N

CAS RN 935-44-4

IUPAC Name: 1-phenylcyclopropane-1-carbonitrile
Synonyms: 1-phenylcyclopropane-1-carbonitrile
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: ZHFURHRJUWYDKG-UHFFFAOYSA-N
SMILES: N#CC1(CC1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)