accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Cl

CAS RN 935-56-8

IUPAC Name: 1-chloroadamantane
Synonyms: 1-chloroadamantane
Molecular Weight (g/mol): 170.68
Molecular Formula: C10H15Cl
InChi Key: OZNXTQSXSHODFR-UHFFFAOYSA-N
SMILES: ClC12CC3CC(CC(C3)C1)C2
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)