accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 936-02-7

IUPAC Name: 2-hydroxybenzohydrazide
Synonyms: benzoylhydrazine, O-hydroxy
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: XSXYESVZDBAKKT-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=CC=C1O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)