accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CHNSHNNH2

CAS RN 93693-01-7

IUPAC Name: 3-amino-1-(4-ethylphenyl)thiourea
Synonyms: 3-amino-1-(4-ethylphenyl)thiourea
Molecular Weight (g/mol): 195.28
Molecular Formula: C9H13N3S
InChi Key: MXVVZBDSLDIVAW-UHFFFAOYSA-N
SMILES: CCC1=CC=C(NC(=S)NN)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results