accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 937-41-7

IUPAC Name: phenyl prop-2-enoate
Synonyms: phenylacrylate
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: WRAQQYDMVSCOTE-UHFFFAOYSA-N
SMILES: C=CC(=O)OC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results