accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 937-52-0

H3CNH2

CAS RN 937-52-0

IUPAC Name: 4-phenylbutan-2-amine
Synonyms: α-benzylisopropylamine benzenepropanamine, �-methyl-
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: WECUIGDEWBNQJJ-UHFFFAOYNA-N
SMILES: CC(N)CCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results