accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N(R)NON(S)(R)HNOHOOOOH3CH3C

CAS RN 93778-57-5

IUPAC Name: 9-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl)-6,9-dihydro-3H-purin-6-one
Synonyms: 9-(5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxyoxolan-2-yl)-3H-purin-6-one
Molecular Weight (g/mol): 554.6
Molecular Formula: C31H30N4O6
InChi Key: IYNGMVFRUKBGNM-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(OCC1OC(CC1O)N1C=NC2=C1NC=NC2=O)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results