accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 939-26-4

Br

CAS RN 939-26-4

IUPAC Name: 2-(bromomethyl)naphthalene
Synonyms: (2-naphthyl)bromomethane
Molecular Weight (g/mol): 221.1
Molecular Formula: C11H9Br
InChi Key: RUHJZSZTSCSTCC-UHFFFAOYSA-N
SMILES: BrCC1=CC=C2C=CC=CC2=C1
Boiling Point 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)