accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOBr

CAS RN 939-54-8

IUPAC Name: 2-bromoethyl benzoate
Synonyms: 2-bromoethyl benzoate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: KNBBDZULQFKSIE-UHFFFAOYSA-N
SMILES: BrCCOC(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)