accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 939759-23-6

IUPAC Name: 2-(bromomethyl)oxetane
Synonyms: 2-(bromomethyl)oxetane
Molecular Weight (g/mol): 151
Molecular Formula: C4H7BrO
InChi Key: OMXAGUVERXNCSZ-UHFFFAOYNA-N
SMILES: BrCC1CCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results