accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOO

CAS RN 94-36-0

IUPAC Name: benzoyl benzenecarboperoxoate
Synonyms: fostex mytolac peroxyde de benzoyle lucidol 50P asidopan diphenylglyoxal peroxide vanoxide aksil 5 dry and clear duresthin 5
Molecular Weight (g/mol): 242.23
Molecular Formula: C14H10O4
InChi Key: OMPJBNCRMGITSC-UHFFFAOYSA-N
SMILES: O=C(OOC(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)