accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 94-52-0

IUPAC Name: 6-nitro-1H-1,3-benzodiazole
Synonyms: 1H-benzimidazole, 6-nitro-
Molecular Weight (g/mol): 163.14
Molecular Formula: C7H5N3O2
InChi Key: XPAZGLFMMUODDK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C2N=CNC2=C1
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results