CAS RN 94-68-8
IUPAC Name:
N-ethyl-2-methylaniline
Synonyms:
benzenamine, N-ethyl-2-methyl-
Molecular Weight (g/mol):
135.21
Molecular Formula:
C9H13N
InChi Key:
MWOUGPLLVVEUMM-UHFFFAOYSA-N
SMILES:
CCNC1=CC=CC=C1C
1
–
1
of
1
results
1
N-Ethyl-o-toluidine(N-Ethyl-2-methyl-benzenamine), TRC
CAS: 94-68-8 Molecular Formula: C9H13N Molecular Weight (g/mol): 135.21 Synonym: Benzenamine, N-ethyl-2-methyl-,o-Toluidine, N-ethyl- (7CI,8CI),N-Ethyl-2-methylbenzenamine,1-(Ethylamino)-2-methylbenzene,2-(Ethylamino)toluene,N-Ethyl-2-methylaniline,N-Ethyl-N-(o-tolyl)amine,N-Ethyl-o-methylaniline,N-Ethyl-o-toluidine,NSC 8888,o-Methyl-N-ethylaniline IUPAC Name: N-ethyl-2-methylaniline SMILES: CCNc1ccccc1C