accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OH2N

CAS RN 94-70-2

IUPAC Name: 2-ethoxyaniline
Synonyms: 2-ethoxyaniline
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: ULHFFAFDSSHFDA-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results