accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOCH3H2N

CAS RN 94-85-9

IUPAC Name: 2,5-diethoxyaniline
Synonyms: 2,5-diethoxyaniline
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: XPKFTIYOZUJAGA-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(OCC)C(N)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results