accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClCl

CAS RN 94-99-5

IUPAC Name: 2,4-dichloro-1-(chloromethyl)benzene
Synonyms: 2,4-dichlorobenzylchloride
Molecular Weight (g/mol): 195.47
Molecular Formula: C7H5Cl3
InChi Key: IRSVDHPYXFLLDS-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(Cl)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)