CAS RN 94035-02-6
IUPAC Name:
5,10,15,20,25,30,35-heptakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34-tetradecaoxaoctacyclo[31.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹]nonatetracontane-36,37,38,39,40,41,42,43,44,45,46,47,48,49-tetradecol; tetrakis(propan-1-ol)
Synonyms:
cyclo-hepta-amylose; tetrakis(propanol)
Molecular Weight (g/mol):
1375.37
Molecular Formula:
C54H102O39
InChi Key:
BZRWCHBEBFPRGF-UHFFFAOYNA-N
SMILES:
CCCO.CCCO.CCCO.CCCO.OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC1C(O)C2O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O
1
–
1
of
1
results
1
Hydroxypropyl β-Cyclodextrin, MP Biomedicals™
CAS: 94035-02-6 Molecular Formula: C54H102O39 Molecular Weight (g/mol): 1375.371 InChI Key: BZRWCHBEBFPRGF-UHFFFAOYSA-N Synonym: 2-hydroxypropy-.beta.-cyclodextrin,2-hydroxypropylether-b-cyclodextrin,hydroxypropyl beta-cyclodextrin,tetrakis 1 propanol ; @cyclo-hepta-amylose,cyclo-hepta-amylose; tetrakis propanol PubChem CID: 44134771 SMILES: CCCO.CCCO.CCCO.CCCO.C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)O)O