accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OON

CAS RN 941-69-5

IUPAC Name: 1-phenyl-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-phenylpyrrole-2,5-dione
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: HIDBROSJWZYGSZ-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)