accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 941-98-0

IUPAC Name: 1-(naphthalen-1-yl)ethan-1-one
Synonyms: 1-acetonaphthone
Molecular Weight (g/mol): 170.21
Molecular Formula: C12H10O
InChi Key: QQLIGMASAVJVON-UHFFFAOYSA-N
SMILES: CC(=O)C1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)