accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOHHO

CAS RN 94105-90-5

IUPAC Name: 3-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-7-ol
Synonyms: equol
Molecular Weight (g/mol): 242.27
Molecular Formula: C15H14O3
InChi Key: ADFCQWZHKCXPAJ-UHFFFAOYNA-N
SMILES: OC1=CC=C(C=C1)C1COC2=C(C1)C=CC(O)=C2
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results