accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 94166-62-8

HHClClH3CONH2H2NN

CAS RN 94166-62-8

IUPAC Name: dihydrogen 6-methoxypyridine-2,3-diamine dichloride
Synonyms: dihydrogen 6-methoxypyridine-2,3-diamine dichloride
Molecular Weight (g/mol): 212.07
Molecular Formula: C6H11Cl2N3O
InChi Key: HFTXXPTXSCLIIP-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].COC1=CC=C(N)C(N)=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results