accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 941716-88-7

IUPAC Name: 2,3,4,5,6-pentafluorophenyl 2-(morpholin-4-yl)-1,3-thiazole-5-carboxylate
Synonyms: 2,3,4,5,6-pentafluorophenyl 2-(morpholin-4-yl)-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 380.29
Molecular Formula: C14H9F5N2O3S
InChi Key: ZFWIQYGUHPHWEU-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(OC(=O)C2=CN=C(S2)N2CCOCC2)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results