accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 941716-89-8

CH3NNONH2

CAS RN 941716-89-8

IUPAC Name: 1-{3-[(6-methylpyrazin-2-yl)oxy]phenyl}methanamine
Synonyms: 1-{3-[(6-methylpyrazin-2-yl)oxy]phenyl}methanamine
Molecular Weight (g/mol): 215.26
Molecular Formula: C12H13N3O
InChi Key: PJVRJIDLKXDROC-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC=CC(CN)=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results